siRNA Details
Gene Symbol: LOC103690019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762747.1 | XP_008760969.1 | 11 | 1112 |
Gene Symbol: Dbf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191748.1 | NP_001178677.1 | 1109 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001190717, NM_013726, AK163408, AJ003132, BC138710, and others.
Gene Ontology Categories:
