siRNA Details
Gene Symbol: Steap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107846.1 | NP_001101316.1 | 3 | 734 | ||
XM_006236032.2 | XP_006236094.1 | 3 | 800 | |||
XM_006236033.2 | XP_006236095.1 | 3 | 1262 | |||
XM_006236034.2 | XP_006236096.1 | 3 | 948 | |||
XM_006236035.1 | XP_006236097.1 | 3 | 689 | |||
XM_006236036.2 | XP_006236098.1 | 3 | 636 | |||
XM_008762703.1 | XP_008760925.1 | 4 | 991 |
