siRNA Details
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107848.1 | NP_001101318.1 | 19 | 1967 | ||
XM_006257072.2 | XP_006257134.1 | 18 | 2408 |

More Information
Other Species Targeted

Gene Ontology Categories:
