siRNA Details
Gene Symbol: Stard8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107849.2 | NP_001101319.2 | 7 | 2115 | ||
XM_006257073.2 | XP_006257135.1 | 8 | 3623 | |||
XM_006257074.2 | XP_006257136.1 | 9 | 2355 | |||
XM_006257075.2 | XP_006257137.1 | 8 | 2239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
