siRNA Details
Gene Symbol: Tmem106b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004267.1 | NP_001004267.1 | 8, 9 | 805 | ||
GenBank mRNA | BC079127.1 | - | 8, 9 | 805 | 7127112 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_027992, BC050246, AK009646, BC022695, AK148477.
Gene Ontology Categories:
