siRNA Details
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034107.1 | NP_001029279.1 | 11 | 1204 | ||
XM_006236140.2 | XP_006236202.1 | 11 | 1246 | |||
XM_006236141.2 | XP_006236203.1 | 10 | 1207 | |||
GenBank mRNA | BC100082.1 | - | 11 | 1204 | 7385392 | |
FQ232896.1 | - | 2 | 120 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023626, AY007791, BC005721, AK146145, BC018342, and others.
Gene Ontology Categories:
