siRNA Details
Gene Symbol: Fam3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198771.1 | NP_942066.1 | 8 | 586 | ||
GenBank mRNA | AY228475.1 | - | 8 | 586 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_138587, BC009086, AY424275, AK004059, BC069880, and others.
Gene Ontology Categories:
