siRNA Details
Gene Symbol: Cadps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749705.3 | XP_003749753.1 | 23, 24 | 3452 | ||
XM_006236152.2 | XP_006236214.1 | 24, 25 | 3572 | |||
XM_006236153.2 | XP_006236215.1 | 24, 25 | 3560 | |||
XM_006236154.2 | XP_006236216.1 | 23, 24 | 3551 | |||
XM_231528.8 | XP_231528.4 | 24, 25 | 3572 |
