siRNA Details
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100708.1 | NP_001094178.1 | 4, 5 | 581 | ||
XM_006236198.2 | XP_006236260.1 | 4, 5 | 629 | |||
XM_006236200.2 | XP_006236262.1 | 4, 5 | 629 | |||
XM_008762772.1 | XP_008760994.1 | 1, 2 | 687 | |||
XM_008762773.1 | XP_008760995.1 | 1, 2 | 686 | |||
XM_008762774.1 | XP_008760996.1 | 4, 5 | 627 | |||
GenBank mRNA | BC161956.1 | - | 4, 5 | 581 | 9027258 |
