siRNA Details
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100708.1 | NP_001094178.1 | 12 | 1785 | ||
XM_008762772.1 | XP_008760994.1 | 9 | 1891 | |||
XM_008762774.1 | XP_008760996.1 | 12 | 1831 | |||
GenBank mRNA | BC161956.1 | - | 12 | 1785 | 9027258 |

More Information
Other Species Targeted

Gene Ontology Categories:
