siRNA Details
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107853.1 | NP_001101323.1 | 5, 6 | 849 | ||
XM_006236314.2 | XP_006236376.1 | 7, 8 | 688 | |||
XM_006236315.2 | XP_006236377.1 | 6, 7 | 1352 | |||
XM_006236316.2 | XP_006236378.1 | 6, 7 | 581 | |||
XM_006236319.2 | XP_006236381.1 | 5, 6 | 1281 | |||
XM_006236321.2 | XP_006236383.1 | 5, 6 | 919 | |||
XM_006236322.2 | XP_006236384.1 | 5, 6 | 534 | |||
XM_006236323.2 | XP_006236385.1 | 7, 8 | 1050 | |||
XM_008762778.1 | XP_008761000.1 | 6, 7 | 3488 | |||
XM_008762779.1 | XP_008761001.1 | 6, 7 | 3487 | |||
XM_008762780.1 | XP_008761002.1 | 6, 7 | 788 | |||
XM_008762781.1 | XP_008761003.1 | 5, 6 | 1301 | |||
GenBank mRNA | BC166855.1 | - | 5, 6 | 870 | 9099059 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_138680, NM_001170849, NM_001170848, BC020058, AK046307, and others.
Gene Ontology Categories:
