siRNA Details
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107853.1 | NP_001101323.1 | 8, 9 | 1142 | ||
XM_006236314.2 | XP_006236376.1 | 10, 11 | 981 | |||
XM_006236315.2 | XP_006236377.1 | 9, 10 | 1645 | |||
XM_006236316.2 | XP_006236378.1 | 9, 10 | 874 | |||
XM_006236319.2 | XP_006236381.1 | 8, 9 | 1574 | |||
XM_006236322.2 | XP_006236384.1 | 8, 9 | 827 | |||
XM_006236323.2 | XP_006236385.1 | 10, 11 | 1343 | |||
XM_008762778.1 | XP_008761000.1 | 9, 10 | 3781 | |||
XM_008762780.1 | XP_008761002.1 | 9, 10 | 1081 | |||
XM_008762781.1 | XP_008761003.1 | 8, 9 | 1594 | |||
GenBank mRNA | BC166855.1 | - | 8, 9 | 1163 | 9099059 |

More Information
Other Species Targeted

Gene Ontology Categories:
