siRNA Details
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107853.1 | NP_001101323.1 | 9 | 1192 | ||
XM_006236314.2 | XP_006236376.1 | 11 | 1031 | |||
XM_006236315.2 | XP_006236377.1 | 10 | 1695 | |||
XM_006236316.2 | XP_006236378.1 | 10 | 924 | |||
XM_006236319.2 | XP_006236381.1 | 9 | 1624 | |||
XM_006236321.2 | XP_006236383.1 | 8 | 1232 | |||
XM_006236322.2 | XP_006236384.1 | 9 | 877 | |||
XM_006236323.2 | XP_006236385.1 | 11 | 1393 | |||
XM_008762778.1 | XP_008761000.1 | 10 | 3831 | |||
XM_008762779.1 | XP_008761001.1 | 9 | 3800 | |||
XM_008762780.1 | XP_008761002.1 | 10 | 1131 | |||
XM_008762781.1 | XP_008761003.1 | 9 | 1644 | |||
GenBank mRNA | BC166855.1 | - | 9 | 1213 | 9099059 |

More Information
Other Species Targeted

Gene Ontology Categories:
