siRNA Details
Gene Symbol: LOC100912416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749737.3 | XP_003749785.1 | 1803 |
Gene Symbol: Fam115c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762926.1 | XP_008761148.1 | 3 | 1802 |
Gene Symbol: LOC100912372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749736.3 | XP_003749784.2 | 1802 |
