siRNA Details
Gene Symbol: Doxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199291.1 | NP_954985.1 | 4 | 2085 | ||
XM_006236462.1 | XP_006236524.1 | 5 | 2111 | |||
GenBank mRNA | BC098900.1 | - | 5 | 2117 | 7443671 |

More Information
Other Species Targeted

Gene Ontology Categories:
