siRNA Details
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749741.3 | XP_003749789.1 | 15 | 1947 | ||
XM_006236474.2 | XP_006236536.1 | 15 | 1821 | |||
XM_006236475.2 | XP_006236537.1 | 13 | 1758 | |||
XM_008762933.1 | XP_008761155.1 | 15 | 1582 | |||
GenBank mRNA | AY325199.1 | - | 5 | 355 | ||
AY724479.1 | - | 15 | 1911 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023670, BC049082, AK088465, AB046173, BC045138, and others.
Gene Ontology Categories:
