siRNA Details
Gene Symbol: Prr15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104527.1 | NP_001097997.1 | 2 | 331 | ||
GenBank mRNA | BC127529.1 | - | 2 | 331 | 7126230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
