siRNA Details
Gene Symbol: Kdm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175764.2 | NP_786940.1 | 11 | 1741 | ||
XM_006236637.2 | XP_006236699.1 | 13 | 2011 | |||
XM_006236639.2 | XP_006236701.1 | 12 | 1968 | |||
GenBank mRNA | FQ222223.1 | - | 12 | 1983 | ||
X59993.1 | - | 11 | 1741 |
