siRNA Details
Gene Symbol: Dysf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107869.1 | NP_001101339.1 | 46 | 5247 | ||
XM_006236758.2 | XP_006236820.1 | 50 | 6266 | |||
XM_006236760.2 | XP_006236822.1 | 49 | 6226 | |||
XM_006236761.2 | XP_006236823.1 | 49 | 5914 | |||
XM_006236764.2 | XP_006236826.1 | 49 | 6202 | |||
XM_006236765.2 | XP_006236827.1 | 49 | 5890 | |||
XM_006236766.2 | XP_006236828.1 | 49 | 6172 | |||
XM_006236767.2 | XP_006236829.1 | 49 | 5860 | |||
XM_006236768.2 | XP_006236830.1 | 48 | 6163 | |||
XM_006236769.2 | XP_006236831.1 | 48 | 5851 | |||
XM_006236770.2 | XP_006236832.1 | 48 | 6133 | |||
XM_006236771.2 | XP_006236833.1 | 48 | 5821 | |||
XM_006236772.2 | XP_006236834.1 | 48 | 6109 | |||
XM_006236773.2 | XP_006236835.1 | 48 | 5797 | |||
XM_006236774.2 | XP_006236836.1 | 47 | 6070 | |||
XM_006236775.2 | XP_006236837.1 | 47 | 5758 | |||
XM_008763067.1 | XP_008761289.1 | 50 | 5953 | |||
XM_008763068.1 | XP_008761290.1 | 46 | 5313 | |||
XM_008763069.1 | XP_008761291.1 | 50 | 6268 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001077694, NM_021469, AF188290, AK087986, BC043692, and others.
Gene Ontology Categories:
