siRNA Details
Gene Symbol: RGD1306746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107872.2 | NP_001101342.2 | 9 | 1735 | ||
XM_008763070.1 | XP_008761292.1 | 9 | 2013 | |||
XM_008763071.1 | XP_008761293.1 | 9 | 1785 | |||
XM_008763072.1 | XP_008761294.1 | 9 | 2135 | |||
XM_008763073.1 | XP_008761295.1 | 9 | 1960 | |||
XM_008763074.1 | XP_008761296.1 | 9 | 3641 | |||
GenBank mRNA | BC166726.1 | - | 9 | 1735 | 7933964 |
