siRNA Details
Gene Symbol: Chchd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013431.1 | NP_001013449.1 | 3 | 312 | ||
GenBank mRNA | BC091407.1 | - | 3 | 312 | 7324948 | |
FQ220478.1 | - | 3 | 177 | |||
FQ220795.1 | - | 3 | 207 | |||
FQ224864.1 | - | 3 | 225 | |||
FQ229282.1 | - | 3 | 177 |
Gene Symbol: LOC100361898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749083.3 | XP_003749131.1 | 1 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
