siRNA Details
Gene Symbol: Trnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024261.2 | NP_001019432.1 | 6 | 867 | ||
XM_008763191.1 | XP_008761413.1 | 6 | 916 | |||
XM_008763192.1 | XP_008761414.1 | 6 | 872 | |||
XM_008763193.1 | XP_008761415.1 | 6 | 826 | |||
GenBank mRNA | BC095841.1 | - | 5 | 792 | 7387807 | |
BC105612.1 | - | 6 | 867 | 7135310 |
