siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107880.1 | NP_001101350.1 | 8 | 889 | ||
XM_006237024.2 | XP_006237086.1 | 8 | 902 | |||
XM_006237025.2 | XP_006237087.1 | 7 | 658 | |||
XM_006237026.1 | XP_006237088.1 | 7 | 534 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026849, BC012436, AK161445, AK181484, AK189716, and others.
Gene Ontology Categories:
