siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107880.1 | NP_001101350.1 | 5 | 623 | ||
XM_006237024.2 | XP_006237086.1 | 5 | 636 | |||
XM_006237025.2 | XP_006237087.1 | 4 | 392 | |||
XM_006237026.1 | XP_006237088.1 | 4 | 268 |
