siRNA Details
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024783.1 | NP_001019954.1 | 3 | 368 | ||
XM_006237027.2 | XP_006237089.1 | 2 | 855 | |||
XM_006237028.2 | XP_006237090.1 | 2 | 343 | |||
GenBank mRNA | BC097951.1 | - | 3 | 368 | 7442421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
