siRNA Details
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007700.1 | NP_001007701.1 | 5 | 660 | ||
XM_008763247.1 | XP_008761469.1 | 5 | 1712 | |||
XM_008763248.1 | XP_008761470.1 | 5 | 725 | |||
GenBank mRNA | BC079226.1 | - | 5 | 660 | 7132318 |
Gene Symbol: LOC100909548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752100.1 | XP_003752148.1 | 5 | 666 | ||
XM_006257201.2 | XP_006257263.1 | 5 | 1712 | |||
XM_006257202.1 | XP_006257264.1 | 5 | 885 |

More Information
Other Species Targeted

Gene Ontology Categories:
