siRNA Details
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007700.1 | NP_001007701.1 | 4 | 512 | ||
XM_008763247.1 | XP_008761469.1 | 4 | 1564 | |||
XM_008763248.1 | XP_008761470.1 | 4 | 577 | |||
GenBank mRNA | BC079226.1 | - | 4 | 512 | 7132318 |
Gene Symbol: LOC100909548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752100.1 | XP_003752148.1 | 4 | 518 | ||
XM_006257201.2 | XP_006257263.1 | 4 | 1564 | |||
XM_006257202.1 | XP_006257264.1 | 4 | 737 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
