siRNA Details
Gene Symbol: Mlf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107889.1 | NP_001101359.1 | 8 | 1197 | ||
XM_006237363.2 | XP_006237425.1 | 9 | 1187 | |||
XM_006237364.2 | XP_006237426.1 | 9 | 1208 |

More Information
Other Species Targeted

Gene Ontology Categories:
