siRNA Details
Gene Symbol: Pianp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014059.1 | NP_001014081.1 | 4 | 658 | ||
XM_006237370.2 | XP_006237432.1 | 4 | 681 | |||
XM_008763285.1 | XP_008761507.1 | 4 | 2433 | |||
XM_008763286.1 | XP_008761508.1 | 4 | 2473 | |||
GenBank mRNA | BC085924.1 | - | 4 | 658 | 7109219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
