siRNA Details
Gene Symbol: Pianp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014059.1 | NP_001014081.1 | 4 | 664 | ||
XM_006237370.2 | XP_006237432.1 | 4 | 687 | |||
XM_008763285.1 | XP_008761507.1 | 4 | 2439 | |||
XM_008763286.1 | XP_008761508.1 | 4 | 2479 | |||
GenBank mRNA | BC085924.1 | - | 4 | 664 | 7109219 |
