siRNA Details
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009419.1 | NP_001009419.1 | 2, 3 | 397 | ||
GenBank mRNA | BC087599.1 | - | 2, 3 | 397 | 7313414 | |
FQ230721.1 | - | 1, 2 | 12 |
