siRNA Details
Gene Symbol: Rassf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191753.1 | NP_001178682.1 | 5 | 1332 | ||
XM_006237659.2 | XP_006237721.1 | 4 | 1251 | |||
XM_006237660.2 | XP_006237722.1 | 5 | 1823 | |||
XM_008763381.1 | XP_008761603.1 | 5 | 3413 |
