siRNA Details
Gene Symbol: Trpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207608.1 | NP_997491.1 | 18 | 2081 | ||
GenBank mRNA | AY496961.1 | - | 18 | 2081 |

More Information
Other Species Targeted

Gene Ontology Categories:
