siRNA Details
Gene Symbol: Prex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107899.1 | NP_001101369.1 | 22 | 2632 | ||
XM_006237753.2 | XP_006237815.1 | 24 | 3223 |

More Information
Other Species Targeted

Gene Ontology Categories:
