siRNA Details
Gene Symbol: Asph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237855.2 | XP_006237917.1 | 18 | 1752 | ||
XM_006237858.2 | XP_006237920.1 | 17 | 1705 | |||
XM_006237859.2 | XP_006237921.1 | 18 | 3036 | |||
XM_006237860.2 | XP_006237922.1 | 17 | 2990 | |||
XM_006237861.2 | XP_006237923.1 | 16 | 1593 | |||
XM_006237862.2 | XP_006237924.1 | 17 | 2923 | |||
XM_008763567.1 | XP_008761789.1 | 19 | 1794 | |||
XM_008763568.1 | XP_008761790.1 | 18 | 1748 | |||
XM_008763569.1 | XP_008761791.1 | 18 | 1750 | |||
XM_008763570.1 | XP_008761792.1 | 19 | 3079 | |||
XM_008763571.1 | XP_008761793.1 | 18 | 1681 | |||
XM_008763572.1 | XP_008761794.1 | 16 | 1661 | |||
XM_008763573.1 | XP_008761795.1 | 18 | 3033 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
