siRNA Details
Gene Symbol: Asph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237855.2 | XP_006237917.1 | 19 | 1917 | ||
XM_006237858.2 | XP_006237920.1 | 18 | 1870 | |||
XM_006237859.2 | XP_006237921.1 | 19 | 3201 | |||
XM_006237860.2 | XP_006237922.1 | 18 | 3155 | |||
XM_006237861.2 | XP_006237923.1 | 17 | 1758 | |||
XM_006237862.2 | XP_006237924.1 | 18 | 3088 | |||
XM_008763567.1 | XP_008761789.1 | 20 | 1959 | |||
XM_008763568.1 | XP_008761790.1 | 19 | 1913 | |||
XM_008763569.1 | XP_008761791.1 | 19 | 1915 | |||
XM_008763570.1 | XP_008761792.1 | 20 | 3244 | |||
XM_008763571.1 | XP_008761793.1 | 19 | 1846 | |||
XM_008763572.1 | XP_008761794.1 | 17 | 1826 | |||
XM_008763573.1 | XP_008761795.1 | 19 | 3198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
