siRNA Details
Gene Symbol: Kdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004268.1 | NP_001004268.1 | 2 | 793 | ||
XM_006239015.2 | XP_006239077.1 | 2 | 1834 | |||
XM_006239016.1 | XP_006239078.1 | 2 | 1018 | |||
XM_006239017.2 | XP_006239079.1 | 2 | 1631 | |||
XM_006239018.2 | XP_006239080.1 | 2 | 924 | |||
XM_008764133.1 | XP_008762355.1 | 1 | 1260 | |||
GenBank mRNA | BC079148.1 | - | 2 | 793 | 7128510 |
Gene Symbol: LOC100911383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773877.1 | XP_008772099.1 | 1018 | |||
XM_008773878.1 | XP_008772100.1 | 1834 | ||||
XM_008773879.1 | XP_008772101.1 | 811 | ||||
XM_008773880.1 | XP_008772102.1 | 1631 | ||||
XM_008773881.1 | XP_008772103.1 | 924 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
