siRNA Details
Gene Symbol: Wdtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107908.1 | NP_001101378.1 | 7 | 1030 | ||
XM_008764134.1 | XP_008762356.1 | 7 | 1169 | |||
XM_008764135.1 | XP_008762357.1 | 7 | 826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
