siRNA Details
Gene Symbol: Ppp1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107911.1 | NP_001101381.1 | 2, 3 | 122 | ||
GenBank mRNA | BC162025.1 | - | 2, 3 | 121 | 9099457 |

More Information
Other Species Targeted

Gene Ontology Categories:
