siRNA Details
Gene Symbol: Ppp1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107911.1 | NP_001101381.1 | 7 | 949 | ||
GenBank mRNA | BC162025.1 | - | 7 | 948 | 9099457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
