siRNA Details
Gene Symbol: Serinc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031656.1 | NP_001026826.1 | 7 | 862 | ||
GenBank mRNA | BC099093.1 | - | 7 | 862 | 7457983 | |
DQ103709.1 | - | 7 | 848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
