siRNA Details
Gene Symbol: Rad54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191755.1 | NP_001178684.1 | 1 | 88 | ||
XM_006237890.2 | XP_006237952.1 | 2 | 1893 | |||
XM_006237892.2 | XP_006237954.1 | 2 | 1893 | |||
XM_008763543.1 | XP_008761765.1 | 2 | 1893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
