siRNA Details
Gene Symbol: Rad54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191755.1 | NP_001178684.1 | 9, 10 | 1758 | ||
XM_006237890.2 | XP_006237952.1 | 10, 11 | 3563 | |||
XM_006237891.1 | XP_006237953.1 | 8, 9 | 3611 | |||
XM_006237892.2 | XP_006237954.1 | 9, 10 | 3272 | |||
XM_008763543.1 | XP_008761765.1 | 10, 11 | 3563 |
