siRNA Details
Gene Symbol: Osgin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237919.2 | XP_006237981.1 | 6 | 1033 | ||
XM_232798.8 | XP_232798.4 | 6 | 863 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Osgin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237919.2 | XP_006237981.1 | 6 | 1033 | ||
XM_232798.8 | XP_232798.4 | 6 | 863 |