siRNA Details
Gene Symbol: Cpne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107917.2 | NP_001101387.2 | 8 | 794 | ||
XM_006237932.1 | XP_006237994.1 | 9 | 916 | |||
GenBank mRNA | AM746508.1 | - | 8 | 794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
