siRNA Details
Gene Symbol: Cpne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107917.2 | NP_001101387.2 | 13 | 1177 | ||
XM_006237932.1 | XP_006237994.1 | 14 | 1299 | |||
GenBank mRNA | AM746508.1 | - | 13 | 1177 |

More Information
Other Species Targeted

Gene Ontology Categories:
