siRNA Details
Gene Symbol: Usp45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107918.1 | NP_001101388.1 | 14 | 1444 | ||
XM_006237938.2 | XP_006238000.1 | 15 | 2129 | |||
XM_006237939.2 | XP_006238001.1 | 15 | 2131 | |||
XM_006237940.2 | XP_006238002.1 | 15 | 2126 | |||
XM_006237941.2 | XP_006238003.1 | 14 | 1985 | |||
GenBank mRNA | FQ227866.1 | - | 1 | 21 |
