siRNA Details
Gene Symbol: Usp45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107918.1 | NP_001101388.1 | 8 | 893 | ||
XM_006237938.2 | XP_006238000.1 | 8 | 921 | |||
XM_006237939.2 | XP_006238001.1 | 8 | 923 | |||
XM_006237940.2 | XP_006238002.1 | 8 | 918 | |||
XM_006237941.2 | XP_006238003.1 | 8 | 921 |

More Information
Other Species Targeted

Gene Ontology Categories:
