siRNA Details
Gene Symbol: Lingo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107926.1 | NP_001101396.1 | 974 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001166001, NM_001166000, NM_001165999, NM_175516, BC137866, and others.
