siRNA Details
Gene Symbol: Abca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178095.2 | NP_835196.1 | 30, 31 | 4405 | ||
XM_008763708.1 | XP_008761930.1 | 33, 34 | 4994 | |||
XM_008763709.1 | XP_008761931.1 | 33, 34 | 4994 | |||
XM_008763710.1 | XP_008761932.1 | 33, 34 | 4987 | |||
GenBank mRNA | AY208182.1 | - | 30, 31 | 4405 |
